2-Bromo-7-iodo-9H-fluoren-9-one 98,0 GC_ANALYSMETOD

Leverantör: TCI
Varning

B5897-25G B5897-5G
TCIAB5897-25GEA 2950 SEK
TCIAB5897-25G TCIAB5897-5G
2-Bromo-7-iodo-9H-fluoren-9-one 98,0 GC_ANALYSMETOD
2-Bromo-7-iodo-9H-fluoren-9-one
Formel: C₁₃H₆BrIO
MW: 385.00 g/mol
MDL Number: MFCD20484974
CAS nummer: 867374-53-6

Order Now

Specifikation testresultat

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 197.0 to 201.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR