tert-Butoxytrimethylsilane 97,0 GC_ANALYSMETOD
Leverantör: TCI
|
Fara
|
B5877-5ML
B5877-25ML
TCIAB5877-5MLEA
670
SEK
TCIAB5877-5ML
TCIAB5877-25ML
tert-Butoxytrimethylsilane 97,0 GC_ANALYSMETOD
tert-Butoxytrimethylsilane
Formel:
C₇H₁₈OSi MW: 146.31 g/mol Kokpunkt: 102°C |
MDL Number:
MFCD00054893 CAS nummer: 13058-24-7 UN: 1993 ADR: 3 II |
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...