3-(3-Bromophenyl)-9-phenyl-9H-carbazole ≥98.0%

Leverantör: TCI EUROPE
Varning

B5869-1G B5869-5G
TCIAB5869-1GEA 1275 SEK
TCIAB5869-1G TCIAB5869-5G
3-(3-Bromophenyl)-9-phenyl-9H-carbazole ≥98.0%
3-(3-Bromophenyl)-9-phenyl-9H-carbazole
Formel: C₂₄H₁₆BrN
MW: 398,30 g/mol
MDL Number: MFCD32062864
CAS nummer: 854952-59-3

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 179.0 to 183.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR