6-Bromo-8-chloroimidazo[1,2-a]pyridine ≥98.0%

Leverantör: TCI EUROPE

B5848-1G B5848-5G
TCIAB5848-1GEA 1150 SEK
TCIAB5848-1G TCIAB5848-5G
6-Bromo-8-chloroimidazo[1,2-a]pyridine ≥98.0%
6-Bromo-8-chloroimidazo[1,2-a]pyridine
Formel: C₇H₄BrClN₂
MW: 231,4800 g/mol
MDL Number: MFCD11518983
CAS nummer: 474708-88-8

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 131.0 to 135.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR