Bis[2-[2-(2-chloroethoxy)ethoxy]ethyl]ether ≥97.0%
Leverantör: TCI
|
Varning
|
B5839-1G
B5839-5G
TCIAB5839-1GEA
910
SEK
TCIAB5839-1G
TCIAB5839-5G
Bis[2-[2-(2-chloroethoxy)ethoxy]ethyl]ether ≥97.0%
Bis[2-[2-(2-chloroethoxy)ethoxy]ethyl]ether
Formel:
C₁₂H₂₄Cl₂O₅ Kokpunkt: 162°C Smältpunkt: -12°C |
MDL Number:
MFCD00068710 CAS nummer: 52559-90-7 |
Specifikation testresultat
Appearance | White to Yellow to Green clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...