1-Bromo-9-phenyl-9H-carbazole ≥97.0%
Leverantör: TCI EUROPE
|
Varning
|
B5811-1G
B5811-5G
TCIAB5811-1GEA
2275
SEK
TCIAB5811-1G
TCIAB5811-5G
1-Bromo-9-phenyl-9H-carbazole ≥97.0%
1-Bromo-9-phenyl-9H-carbazole
Formel:
C₁₈H₁₂BrN MW: 322,21 g/mol |
MDL Number:
MFCD28975084 CAS nummer: 1333002-37-1 |
Specifikation testresultat
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 73.0 to 77.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...