Butyltrimethylammonium bromide ≥97.0%
Leverantör: TCI
|
Varning
|
Synonyms:
N,N,N-Trimethylbutan-1-aminium bromide
B5798-5G
B5798-25G
TCIAB5798-5GEA
670
SEK
TCIAB5798-5G
TCIAB5798-25G
Butyltrimethylammonium bromide ≥97.0%
Butyltrimethylammonium bromide
Formel:
C₇H₁₈BrN Smältpunkt: 196°C |
MDL Number:
MFCD12761450 CAS nummer: 2650-51-3 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (Nonaqueous Titration) | min. 97.0 % |
Purity (Argentmetric Titration) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...