2-(3-Biphenylyl)amino-9,9-dimethylfluorene
Leverantör: TCI EUROPE
Synonyms:
N-([1,1'-Biphenyl]-3-yl)-9,9-dimethyl-9H-fluoren-2-amine
B5793-1G
B5793-5G
TCIAB5793-1GEA
685
SEK
TCIAB5793-1G
TCIAB5793-5G
2-(3-Biphenylyl)amino-9,9-dimethylfluorene
2-(3-Biphenylyl)amino-9,9-dimethylfluorene
Formel:
C₂₇H₂₃N MW: 361,49 g/mol |
MDL Number:
MFCD30471917 CAS nummer: 1372778-66-9 |
Specifikation testresultat
Appearance | White to Green to Brown powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 135.0 to 139.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...