5-Bromo-2-nitrophenol ≥98.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Varning

Synonyms: 1-Bromo-3-hydroxy-4-nitrobenzene

B5788-5G B5788-25G
TCIAB5788-5GEA 965 SEK
TCIAB5788-5G TCIAB5788-25G
5-Bromo-2-nitrophenol ≥98.0% (by GC, titration analysis)
5-Bromo-2-nitrophenol
Formel: C₆H₄BrNO₃
MDL Number: MFCD03095027
CAS nummer: 27684-84-0

Order Now

Specifikation testresultat

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 42.0 to 46.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR