1-Bromodibenzofuran ≥98.0%

Leverantör: TCI EUROPE

Synonyms: 1-Bromodibenzo[b,d]furan

B5766-200MG B5766-1G
TCIAB5766-200MGEA 365 SEK
TCIAB5766-200MG TCIAB5766-1G
1-Bromodibenzofuran ≥98.0%
1-Bromodibenzofuran
Formel: C₁₂H₇BrO
MW: 247,09 g/mol
MDL Number: MFCD00094238
CAS nummer: 50548-45-3

Order Now

Specifikation testresultat

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 65.0 to 69.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR