3-Bromo-4'-chloro-1,1'-biphenyl ≥97.0%

Leverantör: TCI EUROPE

B5764-25G B5764-5G
TCIAB5764-25GEA 3125 SEK
TCIAB5764-25G TCIAB5764-5G
3-Bromo-4'-chloro-1,1'-biphenyl ≥97.0%
3-Bromo-4'-chloro-1,1'-biphenyl
Formel: C₁₂H₈BrCl
MW: 267,55 g/mol
MDL Number: MFCD06201387
CAS nummer: 164334-69-4
UN: 3151
ADR: 9,II

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR