2-Bromo-1,1':4',1''-terphenyl ≥98.0%

Leverantör: TCI EUROPE

Synonyms: 2-Bromo-p-terphenyl

B5760-1G B5760-5G
TCIAB5760-1GEA 1500 SEK
TCIAB5760-1G TCIAB5760-5G
2-Bromo-1,1':4',1''-terphenyl ≥98.0%
2-Bromo-1,1':4',1''-terphenyl
Formel: C₁₈H₁₃Br
MW: 309,21 g/mol
MDL Number: MFCD00093613
CAS nummer: 3282-24-4
UN: 3152
ADR: 9,II

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 85.0 to 89.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR