3-Bromo-3'-chloro-1,1'-biphenyl ≥98.0%

Leverantör: TCI EUROPE

B5759-5G B5759-25G
TCIAB5759-5GEA 655 SEK
TCIAB5759-5G TCIAB5759-25G
3-Bromo-3'-chloro-1,1'-biphenyl ≥98.0%
3-Bromo-3'-chloro-1,1'-biphenyl
Formel: C₁₂H₈BrCl
MW: 267,55 g/mol
MDL Number: MFCD06201361
CAS nummer: 844856-42-4
UN: 3152
ADR: 9,II

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 39.0 to 43.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR