1-(4-Bromophenyl)pyrene ≥98.0%

Leverantör: TCI EUROPE

B5755-1G B5755-5G
TCIAB5755-1GEA 950 SEK
TCIAB5755-1G TCIAB5755-5G
1-(4-Bromophenyl)pyrene ≥98.0%
1-(4-Bromophenyl)pyrene
Formel: C₂₂H₁₃Br
MW: 357,25 g/mol
MDL Number: MFCD30474735
CAS nummer: 345924-29-0

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 151.0 to 155.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR