2-[2-[(tert-Butoxycarbonyl)amino]ethoxy]ethanol ≥98.0%
Leverantör: TCI EUROPE
Synonyms: tert-Butyl [2-(2-Hydroxyethoxy)ethyl]carbamate, tert-Butyl (2-(2-hydroxyethoxy)ethyl)carbamate, [2-(2-Hydroxyethoxy)ethyl]carbamic acid tert-Butyl ester, 2-[2-(Boc-amino)ethoxy]ethanol
B5745-5G B5745-25G
TCIAB5745-5GEA 645 SEK
TCIAB5745-5G TCIAB5745-25G
2-[2-[(tert-Butoxycarbonyl)amino]ethoxy]ethanol ≥98.0%
2-[2-[(tert-Butoxycarbonyl)amino]ethoxy]ethanol
Formel: C₉H₁₉NO₄
MW: 205,25 g/mol
MDL Number: MFCD04038542
CAS nummer: 139115-91-6
Order Now
Specifikation testresultat
Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %