4-Bromo-1,2-diiodobenzene ≥98.0%

Leverantör: TCI EUROPE
Varning

B5743-1G B5743-5G
TCIAB5743-1GEA 1050 SEK
TCIAB5743-1G TCIAB5743-5G
4-Bromo-1,2-diiodobenzene ≥98.0%
4-Bromo-1,2-diiodobenzene
Formel: C₆H₃BrI₂
MW: 408,80 g/mol
MDL Number: MFCD07778994
CAS nummer: 21521-54-0

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 49.0 to 53.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR