4-Bromo-N-methoxy-N-methylbenzamide ≥95.0%
Leverantör: TCI EUROPE
|
Varning
|
B5728-1G
B5728-5G
TCIAB5728-1GEA
615
SEK
TCIAB5728-1G
TCIAB5728-5G
4-Bromo-N-methoxy-N-methylbenzamide ≥95.0%
4-Bromo-N-methoxy-N-methylbenzamide
Formel:
C₉H₁₀BrNO₂ MW: 244,09 g/mol |
MDL Number:
MFCD02684303 CAS nummer: 192436-83-2 |
Specifikation testresultat
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...