5-(Bromomethyl)undecane ≥97.0%

Leverantör: TCI
Varning

B5721-25G B5721-5G
TCIAB5721-25GEA 5825 SEK
TCIAB5721-25G TCIAB5721-5G
5-(Bromomethyl)undecane ≥97.0%
5-(Bromomethyl)undecane
Formel: C₁₂H₂₅Br
Kokpunkt: 85°C
MDL Number: MFCD23115881
CAS nummer: 85531-02-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR