2-Bromo-1-tetralone ≥97.0%

Leverantör: TCI EUROPE

Synonyms: 2-Bromo-3,4-dihydronaphthalen-1(2H)-one, 2-Bromo-3,4-dihydro-1(2H)-naphthalenone

B5698-1G B5698-5G
TCIAB5698-1GEA 910 SEK
TCIAB5698-1G TCIAB5698-5G
2-Bromo-1-tetralone ≥97.0%
2-Bromo-1-tetralone
Formel: C₁₀H₉BrO
MW: 225,08 g/mol
MDL Number: MFCD00089999
CAS nummer: 13672-07-6
EINECS: 237-154-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3f9c475d858a70accd19c6e23f75ced7a48b0a93&vid=3f9c475d858a70accd19c6e23f75ced7a48b0a93&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 97.0 %
Melting point 37.0 to 41.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR