1,3:2,4-Bis-O-(3,4-dimethylbenzylidene)-D-sorbitol ≥98.0%

Leverantör: TCI EUROPE

Synonyms: 1,3:2,4-Bis-O-[(3,4-dimethylphenyl)methylene]-D-glucitol, (1R)-1-((4R,4aR,8aS)-2,6-Bis(3,4-dimethylphenyl)tetrahydro-[1,3]dioxino[5,4-d][1,3]dioxin-4-yl)ethane-1,2-diol, DMDBS

B5693-25G B5693-5G
TCIAB5693-25GEA 855 SEK
TCIAB5693-25G TCIAB5693-5G
1,3:2,4-Bis-O-(3,4-dimethylbenzylidene)-D-sorbitol ≥98.0%
1,3:2,4-Bis-O-(3,4-dimethylbenzylidene)-D-sorbitol
Formel: C₂₄H₃₀O₆
MW: 414,50 g/mol
Smältpunkt: 275°C
MDL Number: MFCD09038711
CAS nummer: 135861-56-2

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Specific rotation [a]20/D +45.0 to +51.0 deg (C=1, pyridine)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR