tert-Butyl (S)-4-(Hydroxymethyl)-2,2-dimethyloxazolidine-3-carboxylate ≥97.0%
Leverantör: TCI
|
Varning
|
Synonyms:
(S)-3-(tert-Butoxycarbonyl)-4-(hydroxymethyl)-2,2-dimethyl-1,3-oxazolidine, (S)-3-Boc-4-(hydroxymethyl)-2,2-dimethyl-1,3-oxazolidine, (S)-4-(Hydroxymethyl)-2,2-dimethyloxazolidine-3-carboxylic acid tert-Butyl ester
B5657-1G
B5657-5G
TCIAB5657-1GEA
1600
SEK
TCIAB5657-1G
TCIAB5657-5G
tert-Butyl (S)-4-(Hydroxymethyl)-2,2-dimethyloxazolidine-3-carboxylate ≥97.0%
tert-Butyl (S)-4-(Hydroxymethyl)-2,2-dimethyloxazolidine-3-carboxylate
Formel:
C₁₁H₂₁NO₄ Storage Temperature: Kylskåp |
MDL Number:
MFCD11041410 CAS nummer: 108149-65-1 |
Specifikation testresultat
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 97.0 % |
Specific rotation [a]20/D | +22.0 to +26.0 deg (C=2, CHCL3) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...