Bis(2,2,6,6-tetramethyl-4-piperidyl-1-oxyl) Sebacate ≥98.0%
Leverantör: TCI EUROPE
Varning
Synonyms: Bis(2,2,6,6-tetramethyl-4-piperidyl-1-oxyl) Decanedioate, Decanedioic acid bis(2,2,6,6-tetramethyl-4-piperidyl-1-oxyl) ester, Sebacic acid bis(2,2,6,6-tetramethyl-4-piperidyl-1-oxyl) ester
B5642-1G B5642-5G
TCIAB5642-1GEA 1225 SEK
TCIAB5642-1G TCIAB5642-5G
Bis(2,2,6,6-tetramethyl-4-piperidyl-1-oxyl) Sebacate ≥98.0%
Bis(2,2,6,6-tetramethyl-4-piperidyl-1-oxyl) Sebacate
Formel: C₂₈H₅₀N₂O₆
MW: 510,72 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD12756000
CAS nummer: 2516-92-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4d2ca4d56a1b2bd19c68ba46936b083d1cc1a53b&vid=4d2ca4d56a1b2bd19c68ba46936b083d1cc1a53b&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Almost white powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (Iodometric Titration) min. 98.0 %
Melting point 99.0 to 103.0 deg-C