6-Bromo-2-chloroquinoline ≥98.0%
Leverantör: TCI EUROPE
Varning
B5628-1G B5628-5G
TCIAB5628-1GEA 805 SEK
TCIAB5628-1G TCIAB5628-5G
6-Bromo-2-chloroquinoline ≥98.0%
6-Bromo-2-chloroquinoline
Formel: C₉H₅BrClN
MW: 242,50 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD04115272
CAS nummer: 1810-71-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d1c19639ef143d32f7ef0e073797b8eefffe651f&vid=d1c19639ef143d32f7ef0e073797b8eefffe651f&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 160.0 to 164.0 deg-C