9-(3-Bromophenyl)-9-phenyl-9H-fluorene ≥98.0%
Leverantör: TCI EUROPE
B5616-1G
B5616-5G
TCIAB5616-1GEA
775
SEK
TCIAB5616-1G
TCIAB5616-5G
9-(3-Bromophenyl)-9-phenyl-9H-fluorene ≥98.0%
9-(3-Bromophenyl)-9-phenyl-9H-fluorene
Formel:
C₂₅H₁₇Br MW: 397,32 g/mol |
MDL Number:
MFCD30187277 CAS nummer: 1257251-75-4 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 130.0 to 135.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...