9-(3-Bromophenyl)-9H-fluoren-9-ol ≥98.0%

Leverantör: TCI EUROPE

Synonyms: 9-(3-Bromophenyl)-9-hydroxy-9H-fluorene

B5615-1G B5615-5G
TCIAB5615-1GEA 1475 SEK
TCIAB5615-1G TCIAB5615-5G
9-(3-Bromophenyl)-9H-fluoren-9-ol ≥98.0%
9-(3-Bromophenyl)-9H-fluoren-9-ol
Formel: C₁₉H₁₃BrO
MW: 337,22 g/mol
MDL Number: MFCD16659044
CAS nummer: 1086641-47-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=796628ec46e852dc69bafa7ba71ba14eaaecdb60&vid=796628ec46e852dc69bafa7ba71ba14eaaecdb60&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 90.0 to 98.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR