2-Bromo-9-(2-ethylhexyl)-9H-carbazole ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
B5611-1G
B5611-5G
TCIAB5611-1GEA
1850
SEK
TCIAB5611-1G
TCIAB5611-5G
2-Bromo-9-(2-ethylhexyl)-9H-carbazole ≥98.0%
2-Bromo-9-(2-ethylhexyl)-9H-carbazole
Formel:
C₂₀H₂₄BrN MW: 358,32 g/mol |
CAS nummer:
856422-39-4 |
Specifikation testresultat
Appearance | Light yellow to Brown clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...