3-Bromo-9-n-octyl-9H-carbazole ≥98.0%

Leverantör: TCI EUROPE
Varning

B5610-1G B5610-5G
TCIAB5610-1GEA 1875 SEK
TCIAB5610-1G TCIAB5610-5G
3-Bromo-9-n-octyl-9H-carbazole ≥98.0%
3-Bromo-9-n-octyl-9H-carbazole
Formel: C₂₀H₂₄BrN
MW: 358,32 g/mol
Kokpunkt: 135°C
MDL Number: MFCD31618112
CAS nummer: 628337-00-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6aa1aebf3ac7cb658a9012fed15d04672744655d&vid=6aa1aebf3ac7cb658a9012fed15d04672744655d&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow to Light red powder to crystal
Purity (GC) min. 98.0 %
Melting point 48.0 to 52.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR