N,N-Bis[(diphenylphosphino)methyl]-3-(triethoxysilyl)propylamine ≥95.0%
Leverantör: TCI
|
Varning
|
B5594-1G
B5594-5G
TCIAB5594-1GEA
975
SEK
TCIAB5594-1G
TCIAB5594-5G
N,N-Bis[(diphenylphosphino)methyl]-3-(triethoxysilyl)propylamine ≥95.0%
N,N-Bis[(diphenylphosphino)methyl]-3-(triethoxysilyl)propylamine
Formel:
C₃₅H₄₅NO₃P₂Si Storage Temperature: Kylskåp |
CAS nummer:
904704-23-0 |
Specifikation testresultat
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (with Total Nitrogen) | 95.0 to 105.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...