1-Boc-4-bromopiperidine ≥98.0% (by GC, titration analysis)
Leverantör: TCI
|
Varning
|
Synonyms:
tert-Butyl 4-Bromopiperidine-1-carboxylate, 4-Bromopiperidine-1-carboxylic acid tert-Butyl ester, 1-Boc-4-bromopiperidine, 1-tert-Butoxycarbonyl-4-bromopiperidine, N-Boc-4-Bromopiperidine
B5585-1G
B5585-5G
TCIAB5585-1GEA
455
SEK
TCIAB5585-1G
TCIAB5585-5G
1-Boc-4-bromopiperidine ≥98.0% (by GC, titration analysis)
1-Boc-4-bromopiperidine
Formel:
C₁₀H₁₈BrNO₂ |
MDL Number:
MFCD04115039 CAS nummer: 180695-79-8 |
Specifikation testresultat
Appearance | White to Light yellow powder to lump |
Purity (GC) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Melting point | 40.0 to 44.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...