1-Butyl-3-methylimidazolium hydrogen sulphate ≥97.0% (by HPLC, total nitrogen)
Leverantör: TCI EUROPE
|
Fara
|
Synonyms:
3-Butyl-1-methyl-1H-imidazol-3-ium hydrogensulphate
B5569-25G
B5569-5G
TCIAB5569-25GEA
1275
SEK
TCIAB5569-25G
TCIAB5569-5G
1-Butyl-3-methylimidazolium hydrogen sulphate ≥97.0% (by HPLC, total nitrogen)
1-Butyl-3-methylimidazolium hydrogen sulphate
Formel:
C₈H₁₆N₂O₄S MW: 236,29 g/mol Flampunkt: 284°C |
MDL Number:
MFCD06798197 CAS nummer: 262297-13-2 UN: 3265 ADR: 8,II |
Specifikation testresultat
Appearance | Colorless to Red to Green clear liquid |
Purity (HPLC) | min. 97.0 area% |
Purity (with Total Nitrogen) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...