1-Butyl-1-methylpyrrolidiniumtrifluormethansulphonate ≥98.0%

Leverantör: TCI
Varning

Synonyms: 1-Butyl-1-methylpyrrolidin-1-ium trifluoromethanesulphonate, 1-Butyl-1-methylpyrrolidinium triflate, 1-Butyl-1-methylpyrrolidinium trifluoromethanesulphonate

B5568-5G B5568-25G
TCIAB5568-5GEA 870 SEK
TCIAB5568-5G TCIAB5568-25G
1-Butyl-1-methylpyrrolidiniumtrifluormethansulphonate ≥98.0%
1-Butyl-1-methylpyrrolidiniumtrifluormethansulphonate
Formel: C₁₀H₂₀F₃NO₃S
MW: 291.33 g/mol
Smältpunkt: 4°C
MDL Number: MFCD08458925
CAS nummer: 367522-96-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a0d96af593df6deac4f4dca0fbf8250c018ef5f3&vid=a0d96af593df6deac4f4dca0fbf8250c018ef5f3&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (with Total Nitrogen) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR