N-(tert-Butoxycarbonyl)-D-threonine methyl ester ≥98.0% (by HPLC, total nitrogen)
Leverantör: TCI EUROPE
Synonyms:
Boc-D-Thr-OMe, N-Boc-D-threonine methyl ester, (2R,3S)-Methyl 2-((tert-butoxycarbonyl)amino)-3-hydroxybutanoate
B5537-1G
B5537-5G
TCIAB5537-1GEA
1450
SEK
TCIAB5537-1G
TCIAB5537-5G
N-(tert-Butoxycarbonyl)-D-threonine methyl ester ≥98.0% (by HPLC, total nitrogen)
N-(tert-Butoxycarbonyl)-D-threonine methyl ester
Formel:
C₁₀H₁₉NO₅ MW: 233,26 g/mol |
MDL Number:
MFCD01073763 CAS nummer: 96099-84-2 |
Specifikation testresultat
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Specific rotation [a]20/D | +18.0 to +22.0 deg (C=0.5, MeOH) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...