Biapenem ≥98.0%
Leverantör: TCI EUROPE
Synonyms: (4R,5S,6S)-3-((6,7-Dihydro-5H-pyrazolo[1,2-a][1,2,4]triazol-4-ium-6-yl)thio)-6-((R)-1-hydroxyethyl)-4-methyl-7-oxo-1-azabicyclo[3.2.0]hept-2-ene-2-carboxylate
B5522-25MG B5522-100MG
TCIAB5522-25MGEA 845 SEK
TCIAB5522-25MG TCIAB5522-100MG
Biapenem ≥98.0%
Biapenem
Formel: C₁₅H₁₈N₄O₄S
MW: 350,39 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD00864863
CAS nummer: 120410-24-4
Merck Index: 14,01197
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eac480f888d74cf43783c682a17ce34a5248a535&vid=eac480f888d74cf43783c682a17ce34a5248a535&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 15.20 to 16.70 %
Specific rotation [a]20/D -30.0 to 335.0 deg (C=0.5, H2O)