(S)-(-)-2-(N-Benzylprolyl)aminobenzophenone ≥98.0%

Leverantör: TCI EUROPE

Synonyms: (S)-N-(2-Benzoylphenyl)-1-benzylpyrrolidine-2-carboxamide

B5519-1G B5519-5G
TCIAB5519-1GEA 765 SEK
TCIAB5519-1G TCIAB5519-5G
(S)-(-)-2-(N-Benzylprolyl)aminobenzophenone ≥98.0%
(S)-(-)-2-(N-Benzylprolyl)aminobenzophenone
Formel: C₂₅H₂₄N₂O₂
MW: 384,470 g/mol
MDL Number: MFCD00145260
CAS nummer: 96293-17-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=23f69d868fe509ad0ed46e7a070e68454ed67095&vid=23f69d868fe509ad0ed46e7a070e68454ed67095&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 97.0 %
Melting point 98.0 to 102.0 deg-C
Specific rotation [a]20/D -136 to -142 deg (C=1,MeOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR