5-Bromobenzoxazole ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
5-Bromobenzo[d]oxazole
B5515-1G
B5515-5G
TCIAB5515-1GEA
565
SEK
TCIAB5515-1G
TCIAB5515-5G
5-Bromobenzoxazole ≥98.0%
5-Bromobenzoxazole
Formel:
C₇H₄BrNO MW: 198,0200 g/mol |
MDL Number:
MFCD03095026 CAS nummer: 132244-31-6 |
Specifikation testresultat
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 36.0 to 40.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...