Bis(diethylamino)dimethylsilane ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: N,N,N',N'-Tetraethyl-1,1-dimethylsilanediamine

B5496-5ML B5496-25ML
TCIAB5496-5MLEA 430 SEK
TCIAB5496-5ML TCIAB5496-25ML
Bis(diethylamino)dimethylsilane ≥98.0%
Bis(diethylamino)dimethylsilane
Formel: C₁₀H₂₆N₂Si
MW: 202,42 g/mol
Kokpunkt: 193°C
Flampunkt: 35°C
MDL Number: MFCD00039879
CAS nummer: 4669-59-4
EINECS: 225-116-0
UN: 1993
ADR: 3,III

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR