4-(tert-Butyl)-2-chloropyridine ≥98.0%

Leverantör: TCI
Varning

B5466-1G B5466-5G
TCIAB5466-1GEA 1350 SEK
TCIAB5466-1G TCIAB5466-5G
4-(tert-Butyl)-2-chloropyridine ≥98.0%
4-(tert-Butyl)-2-chloropyridine
Formel: C₉H₁₂ClN
MW: 169.65 g/mol
MDL Number: MFCD12131129
CAS nummer: 81167-60-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=def2b5eaed356b47f80b0e7aaea3d13692c3f325&vid=def2b5eaed356b47f80b0e7aaea3d13692c3f325&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR