9-Benzyl-2,7-dibromo-9H-carbazole ≥98.0% (by GC, total nitrogen)

Leverantör: TCI EUROPE

B5462-1G B5462-5G
TCIAB5462-1GEA 1525 SEK
TCIAB5462-1G TCIAB5462-5G
9-Benzyl-2,7-dibromo-9H-carbazole ≥98.0% (by GC, total nitrogen)
9-Benzyl-2,7-dibromo-9H-carbazole
Formel: C₁₉H₁₃Br₂N
MW: 415,13 g/mol
MDL Number: MFCD30469065
CAS nummer: 1384281-49-5

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (with Total Nitrogen) min. 95.0 %
Melting point 161.0 to 165.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR