3-Bromodibenzothiophene ≥98.0%

Leverantör: TCI EUROPE

Synonyms: 3-Bromodibenzo[b,d]thiophene

B5452-1G B5452-5G
TCIAB5452-1GEA 925 SEK
TCIAB5452-1G TCIAB5452-5G
3-Bromodibenzothiophene ≥98.0%
3-Bromodibenzothiophene
Formel: C₁₂H₇BrS
MW: 263,15 g/mol
MDL Number: MFCD11052997
CAS nummer: 97511-04-1

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 97.0 to 101.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR