3-Bromodibenzofuran ≥98.0%

Leverantör: TCI EUROPE

Synonyms: 3-Bromodibenzo[b,d]furan

B5451-200MG B5451-1G
TCIAB5451-200MGEA 295 SEK
TCIAB5451-200MG TCIAB5451-1G
3-Bromodibenzofuran ≥98.0%
3-Bromodibenzofuran
Formel: C₁₂H₇BrO
MW: 247,09 g/mol
MDL Number: MFCD09966042
CAS nummer: 26608-06-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=163e7d8b4be8eb4bf20268dacbd57a8aac201614&vid=163e7d8b4be8eb4bf20268dacbd57a8aac201614&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 121.0 to 125.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR