Benzyl(4-hydroxyphenyl)methylsulphonium Hexafluoroantimonate ≥98.0% (by HPLC, titration analysis)

Leverantör: TCI EUROPE
Varning

B5428-25G B5428-5G
TCIAB5428-25GEA 5000 SEK
TCIAB5428-25G TCIAB5428-5G
Benzyl(4-hydroxyphenyl)methylsulphonium Hexafluoroantimonate ≥98.0% (by HPLC, titration analysis)
Benzyl(4-hydroxyphenyl)methylsulphonium Hexafluoroantimonate
Formel: C₁₄H₁₅F₆OSSb
MW: 467,08 g/mol
CAS nummer: 125662-42-2
UN: 3077
ADR: 9,III

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Ion exchange titration) min. 98.0 %
Melting point 115.0 to 120.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR