N4-(3-Bromophenyl)quinazoline-4,6-diamine ≥98.0%

Leverantör: TCI EUROPE
Varning

B5416-25MG B5416-100MG
TCIAB5416-25MGEA 790 SEK
TCIAB5416-25MG TCIAB5416-100MG
N4-(3-Bromophenyl)quinazoline-4,6-diamine ≥98.0%
N4-(3-Bromophenyl)quinazoline-4,6-diamine
Formel: C₁₄H₁₁BrN₄
Storage Temperature: Kylskåp
MDL Number: MFCD09908082
CAS nummer: 169205-78-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=061df60d226fed01da81536a56d284e2188400e6&vid=061df60d226fed01da81536a56d284e2188400e6&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 16.50 to 18.50 %
Melting point 202.0 to 206.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR