5-Bromo-4-chloro-3-indolyl-β-D-glucoside ≥98.0%
Leverantör: TCI EUROPE
Synonyms: 5-Bromo-4-chloro-3-indolyl beta-D-Glucopyranoside, (2S,3R,4S,5S,6R)-2-((5-Bromo-4-chloro-1H-indol-3-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol, X-Glc, 5-Bromo-4-chloro-3-(beta-D-glucopyranosyloxy)indole
B5393-200MG B5393-1G
TCIAB5393-200MGEA 350 SEK
TCIAB5393-200MG TCIAB5393-1G
5-Bromo-4-chloro-3-indolyl-β-D-glucoside ≥98.0%
5-Bromo-4-chloro-3-indolyl-beta-D-glucoside
Formel: C₁₄H₁₅BrClNO₆
MW: 408,629 g/mol
Smältpunkt: 243°C
Storage Temperature: Frys
MDL Number: MFCD00063690
CAS nummer: 15548-60-4
EINECS: 239-603-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f2561291d88630e9d9ed900dc75edd8420f498b5&vid=f2561291d88630e9d9ed900dc75edd8420f498b5&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 97.0 %
Specific rotation [a]20/D -78.0 to -83.0 deg (C=0.5,DMF)