tert-Butyl-6-oxa-3-azabicyclo[3.1.0]hexane-3-carboxylate ≥95.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 6-Oxa-3-azabicyclo[3.1.0]hexane-3-carboxylic acid tert-Butyl ester, tert-Butyl 6-oxa-3-azabicyclo[3.1.0]hexane-3-carboxylate, 1-(tert-Butoxycarbonyl)-3,4-epoxypyrrolidine, 1-Boc-3,4-epoxypyrrolidine

B5385-1G B5385-5G
TCIAB5385-1GEA 430 SEK
TCIAB5385-1G TCIAB5385-5G
tert-Butyl-6-oxa-3-azabicyclo[3.1.0]hexane-3-carboxylate ≥95.0%
tert-Butyl-6-oxa-3-azabicyclo[3.1.0]hexane-3-carboxylate
Formel: C₉H₁₅NO₃
MW: 185,2203 g/mol
MDL Number: MFCD08691407
CAS nummer: 114214-49-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1391c858553656c7a622cb02bb618f900e94c12b&vid=1391c858553656c7a622cb02bb618f900e94c12b&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Orange to Green clear liquid
Purity (GC) min. 95.0 %
Acid Value max. 3.0

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR