(3-Bromopropoxy)-tert-butyldimethylsilane ≥98.0%
Leverantör: TCI EUROPE
Varning
Synonyms: (3-Bromopropoxy)(tert-butyl)dimethylsilane
B5346-5G B5346-25G
TCIAB5346-5GEA 925 SEK
TCIAB5346-5G TCIAB5346-25G
(3-Bromopropoxy)-tert-butyldimethylsilane ≥98.0%
(3-Bromopropoxy)-tert-butyldimethylsilane
Formel: C₉H₂₁BrOSi
MW: 253,26 g/mol
Kokpunkt: 90°C
Flampunkt: 85°C
Storage Temperature: Kylskåp
MDL Number: MFCD00216589
CAS nummer: 89031-84-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5df527d307a55bba78d02bf12fd419736c547a02&vid=5df527d307a55bba78d02bf12fd419736c547a02&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %