2-Benzoyl-5-bromothiophene ≥98.0%
Leverantör: TCI EUROPE
Synonyms:
(5-Bromothiophen-2-yl)(phenyl)methanone
B5340-1G
B5340-5G
TCIAB5340-1GEA
655
SEK
TCIAB5340-1G
TCIAB5340-5G
2-Benzoyl-5-bromothiophene ≥98.0%
2-Benzoyl-5-bromothiophene
Formel:
C₁₁H₇BrOS MW: 267,14 g/mol |
MDL Number:
MFCD03086026 CAS nummer: 31161-46-3 |
Specifikation testresultat
Appearance | White to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 72.0 to 76.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...