2-Bromo-9,9-difluoro-7-iodo-9H-fluorene ≥96.0%

Leverantör: TCI EUROPE
Varning

B5327-200MG B5327-1G
TCIAB5327-200MGEA 815 SEK
TCIAB5327-200MG TCIAB5327-1G
2-Bromo-9,9-difluoro-7-iodo-9H-fluorene ≥96.0%
2-Bromo-9,9-difluoro-7-iodo-9H-fluorene
Formel: C₁₃H₆BrF₂I
MW: 407,00 g/mol
CAS nummer: 1499193-60-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6dae8e13f7d9e2bdad7815ef277cea393d10e50f&vid=6dae8e13f7d9e2bdad7815ef277cea393d10e50f&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 96.5 %
Melting point 141.0 to 145.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR