1H-(N-Benzyl)-4-iodopyrazole ≥98.0%

Leverantör: TCI EUROPE
Fara

Synonyms: 1-Benzyl-4-iodo-1H-pyrazole

B5286-1G B5286-5G
TCIAB5286-1GEA 815 SEK
TCIAB5286-1G TCIAB5286-5G
1H-(N-Benzyl)-4-iodopyrazole ≥98.0%
1H-(N-Benzyl)-4-iodopyrazole
Formel: C₁₀H₉IN₂
MW: 284,1 g/mol
MDL Number: MFCD04038745
CAS nummer: 50877-42-4

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 61.0 to 65.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR