2-(4-Bromophenyl)propan-2-ol ≥98.0%
Leverantör: TCI EUROPE
Synonyms: 2-(4-Bromophenyl)-2-propanol
B5274-1G B5274-5G
TCIAB5274-1GEA 1150 SEK
TCIAB5274-1G TCIAB5274-5G
2-(4-Bromophenyl)propan-2-ol ≥98.0%
2-(4-Bromophenyl)propan-2-ol
Formel: C₉H₁₁BrO
Kokpunkt: 85°C
Storage Temperature: Kylskåp
MDL Number: MFCD11870096
CAS nummer: 2077-19-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4b87ace876aa95a721d8544b31977d310f9e52b0&vid=4b87ace876aa95a721d8544b31977d310f9e52b0&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Orange to Green powder to lump
Purity (GC) min. 98.0 %
Melting point 42.0 to 46.0 deg-C