2-Bromodibenzothiophene 5,5-Dioxide ≥95.0%
Leverantör: TCI EUROPE
Synonyms: 2-Bromodibenzothiophene 5,5-dioxide, 2-Bromodibenzosulpholane, 2-Bromodibenzothiophene sulphone
B5267-200MG B5267-1G
TCIAB5267-200MGEA 1375 SEK
TCIAB5267-200MG TCIAB5267-1G
2-Bromodibenzothiophene 5,5-Dioxide ≥95.0%
2-Bromodibenzothiophene 5,5-Dioxide
Formel: C₁₂H₇BrO₂S
MW: 295,15 g/mol
Smältpunkt: 267°C
MDL Number: MFCD00623266
CAS nummer: 53846-85-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c144df98f9bf284db968ab68d30e0b124168bd29&vid=c144df98f9bf284db968ab68d30e0b124168bd29&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Light yellow powder to crystal
Purity (GC) min. 95.0 %